Search
Search

ARFIP1| Gene Symbol: ARFIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252398.1 | NP_001239327.1 | 4 - 5 | 394 | 79 | |
| XM_015276326.2 | XP_015131812.1 | 3 - 4 | 304 | 79 | ||
| XM_015276329.2 | XP_015131815.1 | 3 - 4 | 380 | 79 | ||
| XM_015276330.2 | XP_015131816.1 | 3 - 4 | 245 | 79 | ||
| XM_025149759.1 | XP_025005527.1 | 5 - 6 | 581 | 79 | ||
Set Membership: |
|