Search
Search

PLXNB3| Gene Symbol: PLXNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257265.1 | NP_001244194.1 | 14 - 15 | 2835 | 69 | |
| XM_025154397.1 | XP_025010165.1 | 17 - 18 | 3980 | 69 | ||
| XM_025154398.1 | XP_025010166.1 | 17 - 18 | 4104 | 69 | ||
Set Membership: |
|