Search
Search

ADAMTS9| Gene Symbol: ADAMTS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004944664.3 | XP_004944721.2 | 40 | 9023 | 89 | |
| XM_004944666.3 | XP_004944723.2 | 39 | 8855 | 89 | ||
| XM_015293287.2 | XP_015148773.1 | 38 | 6524 | 89 | ||
| XM_414417.5 | XP_414417.3 | 40 | 9020 | 89 | ||
| GenBank mRNA | CR386892.1 | - | 1 | 1020 | 89 | |
Set Membership: |
|