Search
Search

GALNT5| Gene Symbol: GALNT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025152501.1 | XP_025008269.1 | 9 - 10 | 1802 | 58 | |
| XM_422169.6 | XP_422169.5 | 8 - 9 | 2110 | 58 | ||
| GenBank mRNA | CR524023.1 | - | 1 - 2 | 318 | 58 | |
Set Membership: |
|