Search
Search

NLRC3| Gene Symbol: NLRC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294675.2 | XP_015150161.2 | 19 | 5471 | 80 | |
| XM_015294676.2 | XP_015150162.2 | 19 | 5359 | 80 | ||
| XM_015294677.2 | XP_015150163.2 | 18 | 5387 | 80 | ||
Set Membership: |
|