Search
Search

TMEM87B| Gene Symbol: TMEM87B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004941650.3 | XP_004941707.3 | 17 - 18 | 1620 | 100 | |
| XM_421155.6 | XP_421155.4 | 17 - 18 | 1617 | 100 | ||
Set Membership: |
|