Search
Search

ASCC1| Gene Symbol: ASCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004942039.3 | XP_004942096.2 | 4 - 5 | 802 | 74 | |
| XM_025151713.1 | XP_025007481.1 | 4 - 5 | 491 | 74 | ||
| XM_025151714.1 | XP_025007482.1 | 4 - 5 | 832 | 74 | ||
| XM_421588.6 | XP_421588.5 | 4 - 5 | 362 | 74 | ||
Set Membership: |
|