Search
Search

MYOF| Gene Symbol: MYOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025151881.1 | XP_025007649.1 | 53 - 54 | 6563 | 81 | |
| XM_025151882.1 | XP_025007650.1 | 53 - 54 | 6561 | 81 | ||
| XM_025151883.1 | XP_025007651.1 | 52 - 53 | 6525 | 81 | ||
| XM_025151884.1 | XP_025007652.1 | 52 - 53 | 6522 | 81 | ||
Set Membership: |
|