Search
Search

CDH12| Gene Symbol: CDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025147686.1 | XP_025003454.1 | 8 - 9 | 2638 | 92 | |
| XM_025147687.1 | XP_025003455.1 | 4 - 5 | 1898 | 92 | ||
| XM_025147688.1 | XP_025003456.1 | 5 - 6 | 1805 | 92 | ||
| XM_025147689.1 | XP_025003457.1 | 6 - 7 | 1228 | 92 | ||
| XM_025147690.1 | XP_025003458.1 | 7 - 8 | 2218 | 92 | ||
| XM_025147691.1 | XP_025003459.1 | 5 - 6 | 1115 | 92 | ||
| XM_025147692.1 | XP_025003460.1 | 5 - 6 | 892 | 92 | ||
Set Membership: |
|