Search
Search

SERPING1| Gene Symbol: SERPING1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025151166.1 | XP_025006934.1 | 7 - 8 | 1368 | 61 | |
| XM_025151167.1 | XP_025006935.1 | 7 - 8 | 1476 | 61 | ||
Set Membership: |
|