Search
Search

SLC7A10| Gene Symbol: SLC7A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154310.1 | XP_025010078.1 | 2 - 3 | 358 | 83 | |
| XM_025154311.1 | XP_025010079.1 | 2 - 3 | 358 | 83 | ||
| XM_414136.6 | XP_414136.3 | 2 - 3 | 358 | 83 | ||
Set Membership: |
|