Search
Search

GPC5| Gene Symbol: GPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938584.3 | XP_004938641.1 | 3 - 4 | 5611 | 91 | |
| XM_004938587.3 | XP_004938644.1 | 3 - 4 | 5610 | 91 | ||
| XM_004938598.3 | XP_004938655.1 | 3 - 4 | 5610 | 91 | ||
| XM_004938599.3 | XP_004938656.1 | 3 - 4 | 5610 | 91 | ||
| XM_004938602.3 | XP_004938659.1 | 3 - 4 | 5609 | 91 | ||
| XM_015274639.2 | XP_015130125.1 | 3 - 4 | 5612 | 91 | ||
| XM_015274645.2 | XP_015130131.1 | 3 - 4 | 1841 | 91 | ||
| XM_015274655.2 | XP_015130141.1 | 3 - 4 | 5610 | 91 | ||
| XM_025146758.1 | XP_025002526.1 | 4 - 5 | 1177 | 91 | ||
| XM_025146759.1 | XP_025002527.1 | 3 - 4 | 5610 | 91 | ||
| XM_025146760.1 | XP_025002528.1 | 3 - 4 | 5610 | 91 | ||
| XM_025146761.1 | XP_025002529.1 | 3 - 4 | 5610 | 91 | ||
| XM_025146762.1 | XP_025002530.1 | 3 - 4 | 5610 | 91 | ||
| XM_025146763.1 | XP_025002531.1 | 3 - 4 | 5610 | 91 | ||
| XM_025146764.1 | XP_025002532.1 | 3 - 4 | 5610 | 91 | ||
| GenBank mRNA | BX933974.1 | - | 3 - 4 | 1025 | 91 | |
Set Membership: |
|