Search
Search

PITPNC1| Gene Symbol: PITPNC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015279977.2 | XP_015135463.1 | 1 - 2 | 653 | 139 | |
| XM_015279981.2 | XP_015135467.1 | 1 - 2 | 656 | 139 | ||
| XM_415676.6 | XP_415676.5 | 1 - 2 | 654 | 139 | ||
| GenBank mRNA | BX934134.2 | - | 1 - 2 | 497 | 139 | |
| CR353717.1 | - | 1 - 2 | 291 | 139 | ||
Set Membership: |
|