Search
Search

ROBO3| Gene Symbol: ROBO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298078.2 | XP_015153564.1 | 4 - 5 | 956 | 115 | |
| XM_015298079.2 | XP_015153565.1 | 4 - 5 | 751 | 115 | ||
| XM_015298080.2 | XP_015153566.1 | 4 - 5 | 751 | 115 | ||
| XM_015298081.2 | XP_015153567.1 | 4 - 5 | 987 | 115 | ||
| XM_025143241.1 | XP_024999009.1 | 4 - 5 | 987 | 115 | ||
Set Membership: |
|