Search
Search

OTOGL| Gene Symbol: OTOGL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015283290.2 | XP_015138776.1 | 53 - 54 | 7580 | 67 | |
| XM_015283295.2 | XP_015138781.1 | 53 - 54 | 7563 | 67 | ||
| XM_015283301.2 | XP_015138787.1 | 53 - 54 | 7580 | 67 | ||
| XM_015283307.2 | XP_015138793.1 | 53 - 54 | 7580 | 67 | ||
| XM_025154451.1 | XP_025010219.1 | 52 - 53 | 7526 | 67 | ||
Set Membership: |
|