Search
Search

CARMIL3| Gene Symbol: CARMIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015272966.2 | XP_015128452.2 | 3 - 4 | 312 | 98 | |
| XM_015272967.2 | XP_015128453.2 | 3 - 4 | 312 | 98 | ||
| XM_015272969.2 | XP_015128455.2 | 3 - 4 | 313 | 98 | ||
| XM_025152651.1 | XP_025008419.1 | 3 - 4 | 312 | 98 | ||
| XM_025152656.1 | XP_025008424.1 | 3 - 4 | 312 | 98 | ||
| XM_025152658.1 | XP_025008426.1 | 3 - 4 | 312 | 98 | ||
| XM_025152660.1 | XP_025008428.1 | 3 - 4 | 312 | 98 | ||
| XM_025152662.1 | XP_025008430.1 | 3 - 4 | 312 | 98 | ||
| XM_025152668.1 | XP_025008436.1 | 3 - 4 | 312 | 98 | ||
| XM_025152673.1 | XP_025008441.1 | 3 - 4 | 312 | 98 | ||
| XM_025152677.1 | XP_025008445.1 | 3 - 4 | 312 | 98 | ||
| XM_025152681.1 | XP_025008449.1 | 3 - 4 | 312 | 98 | ||
| XM_025152683.1 | XP_025008451.1 | 3 - 4 | 312 | 98 | ||
| XM_025152694.1 | XP_025008462.1 | 3 - 4 | 312 | 98 | ||
| XM_025152696.1 | XP_025008464.1 | 3 - 4 | 313 | 98 | ||
Set Membership: |
|