Search
Search

RNF17| Gene Symbol: RNF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015279441.2 | XP_015134927.1 | 19 - 20 | 2999 | 89 | |
| XM_015279446.2 | XP_015134932.1 | 19 - 20 | 2999 | 89 | ||
Set Membership: |
|