Search
Search

TBC1D32| Gene Symbol: TBC1D32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004940260.2 | XP_004940317.1 | 21 - 22 | 2567 | 87 | |
| XM_004940263.3 | XP_004940320.1 | 21 - 22 | 2567 | 87 | ||
| XM_025149167.1 | XP_025004935.1 | 21 - 22 | 2567 | 87 | ||
| XM_025149168.1 | XP_025004936.1 | 20 - 21 | 2507 | 87 | ||
| XM_025149169.1 | XP_025004937.1 | 21 - 22 | 2567 | 87 | ||
Set Membership: |
|