Search
Search

WIPF2| Gene Symbol: WIPF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015299559.2 | XP_015155045.2 | 2 - 3 | 214 | 114 | |
| XM_025144193.1 | XP_024999961.1 | 2 - 3 | 214 | 114 | ||
| XM_025144194.1 | XP_024999962.1 | 2 - 3 | 658 | 114 | ||
Set Membership: |
|