Search
Search

ABCA7| Gene Symbol: ABCA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015299986.2 | XP_015155472.1 | 11 - 12 | 2237 | 66 | |
| XM_025144353.1 | XP_025000121.1 | 11 - 12 | 2359 | 66 | ||
| XM_025144354.1 | XP_025000122.1 | 11 - 12 | 2352 | 66 | ||
Set Membership: |
|