Search
Search

WRKY1| Gene Symbol: WRKY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249973.1 | NP_001236902.1 | 2 - 3 | 671 | 76 | |
| GenBank mRNA | AY323128.1 | - | 2 - 3 | 1135 | 76 | |
| EU019555.1 | - | 2 - 3 | 671 | 76 | ||
| EU375350.1 | - | 2 - 3 | 702 | 76 | ||
Set Membership: |
|