Search
Search

DGAT| Gene Symbol: DGAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255528.1 | NP_001242457.1 | 4 - 5 | 585 | 88 | |
| GenBank mRNA | AY652765.1 | - | 3 - 4 | 247 | 88 | |
| HQ636610.1 | - | 4 - 5 | 585 | 88 | ||
Set Membership: |
|