Search
Search

ALAAT1| Gene Symbol: ALAAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250624.1 | NP_001237553.1 | 9 - 10 | 833 | 107 | |
| GenBank mRNA | EU165371.1 | - | 9 - 10 | 833 | 107 | |
Set Membership: |
|