Search
Search

LOC100101846
LOC547638| Gene Symbol: LOC100101846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF345652.1 | - | 2 | 1453 | 95 | |
| Gene Symbol: LOC547638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY308652.1 | - | 2 | 779 | 95 | |
| AY308653.1 | - | 2 | 958 | 95 | ||
| AY308654.1 | - | 2 | 1289 | 95 | ||
Set Membership: |
|