Search
Search

LOC100794003| Gene Symbol: LOC100794003 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254104.2 | NP_001241033.1 | 5 - 6 | 760 | 70 | |
| GenBank mRNA | BT097204.1 | - | 5 - 6 | 764 | 70 | |
Set Membership: |
|