Search
Search

LOC100810870| Gene Symbol: LOC100810870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255420.2 | NP_001242349.1 | 1 - 2 | 54 | 64 | |
| GenBank mRNA | BT097450.1 | - | 1 - 2 | 56 | 64 | |
Set Membership: |
|