Search
Search

LOC100809900| Gene Symbol: LOC100809900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255648.2 | NP_001242577.1 | 2 - 3 | 410 | 59 | |
| GenBank mRNA | BT093466.1 | - | 2 - 3 | 412 | 59 | |
Set Membership: |
|