Search
Search

LOC100786716| Gene Symbol: LOC100786716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252845.2 | NP_001239774.1 | 6 | 1124 | 97 | |
| GenBank mRNA | BT097572.1 | - | 1 | 61 | 97 | |
| BT097578.1 | - | 1 | 61 | 97 | ||
Set Membership: |
|