Search
Search

SLR1| Gene Symbol: SLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253284.2 | NP_001240213.1 | 1 | 221 | 98 | |
| GenBank mRNA | AK287398.1 | - | 1 | 172 | 98 | |
| FJ534642.1 | - | 1 | 226 | 98 | ||
Set Membership: |
|