Search
Search

Fermt2| Gene Symbol: Fermt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835521.3 | XP_004835578.1 | 11 - 12 | 1418 | 69 | |
| XM_004835522.3 | XP_004835579.1 | 10 - 11 | 1385 | 69 | ||
| XM_004835523.3 | XP_004835580.1 | 10 - 11 | 1385 | 69 | ||
| XM_013074712.2 | XP_012930166.1 | 11 - 12 | 1483 | 69 | ||
| XM_013074714.2 | XP_012930168.1 | 11 - 12 | 1418 | 69 | ||
| XM_013074717.2 | XP_012930171.1 | 10 - 11 | 1385 | 69 | ||
Set Membership: |
|