Search
Search

Fbxl18| Gene Symbol: Fbxl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840382.3 | XP_004840439.1 | 7 - 8 | 2432 | 82 | |
| XM_004840383.2 | XP_004840440.1 | 5 - 6 | 2326 | 82 | ||
| XM_021254648.1 | XP_021110307.1 | 7 - 8 | 2474 | 82 | ||
| XM_021254649.1 | XP_021110308.1 | 6 - 7 | 2411 | 82 | ||
| XM_021254650.1 | XP_021110309.1 | 5 - 6 | 2263 | 82 | ||
| XM_021254651.1 | XP_021110310.1 | 5 - 6 | 2262 | 82 | ||
| XM_021254652.1 | XP_021110311.1 | 5 - 6 | 2277 | 82 | ||
Set Membership: |
|