Search
Search

Thap9| Gene Symbol: Thap9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847962.3 | XP_004848019.1 | 2 - 3 | 420 | 66 | |
| XM_021260962.1 | XP_021116621.1 | 2 - 3 | 426 | 66 | ||
| XM_021260963.1 | XP_021116622.1 | 2 - 3 | 364 | 66 | ||
| XM_021260964.1 | XP_021116623.1 | 2 - 3 | 306 | 66 | ||
| XM_021260965.1 | XP_021116624.1 | 2 - 3 | 426 | 66 | ||
Set Membership: |
|