Search
Search

Ppp2r5c| Gene Symbol: Ppp2r5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836936.3 | XP_004836993.1 | 12 - 13 | 1445 | 64 | |
| XM_004836937.3 | XP_004836994.1 | 12 - 13 | 1466 | 64 | ||
| XM_004836938.3 | XP_004836995.1 | 10 - 11 | 1279 | 64 | ||
| XM_004836940.3 | XP_004836997.1 | 10 - 11 | 1277 | 64 | ||
| XM_013066469.2 | XP_012921923.1 | 10 - 11 | 1321 | 64 | ||
| XM_021244143.1 | XP_021099802.1 | 12 - 13 | 1474 | 64 | ||
| XM_021244159.1 | XP_021099818.1 | 12 - 13 | 1303 | 64 | ||
Set Membership: |
|