Search
Search

Znf84| Gene Symbol: Znf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844046.2 | XP_004844103.1 | 2 - 3 | 418 | 87 | |
| XM_004844048.3 | XP_004844105.1 | 3 - 4 | 1088 | 87 | ||
| XM_004844049.2 | XP_004844106.1 | 3 - 4 | 1094 | 87 | ||
| XM_004844050.2 | XP_004844107.1 | 4 - 5 | 739 | 87 | ||
| XM_004844051.2 | XP_004844108.1 | 3 - 4 | 564 | 87 | ||
Set Membership: |
|