Search
Search

Enkd1| Gene Symbol: Enkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256856.1 | XP_021112515.1 | 6 - 7 | 979 | 67 | |
| XM_021256857.1 | XP_021112516.1 | 6 - 7 | 937 | 67 | ||
| XM_021256858.1 | XP_021112517.1 | 6 - 7 | 970 | 67 | ||
| XM_021256859.1 | XP_021112518.1 | 6 - 7 | 895 | 67 | ||
Set Membership: |
|