Search
Search

Tmem2| Gene Symbol: Tmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838137.3 | XP_004838194.1 | 20 - 21 | 3688 | 62 | |
| XM_004838138.3 | XP_004838195.1 | 20 - 21 | 3678 | 62 | ||
| XM_004838139.3 | XP_004838196.1 | 20 - 21 | 3751 | 62 | ||
| XM_004838140.3 | XP_004838197.1 | 20 - 21 | 3693 | 62 | ||
| XM_004838141.3 | XP_004838198.1 | 20 - 21 | 3674 | 62 | ||
| XM_021252717.1 | XP_021108376.1 | 19 - 20 | 3596 | 62 | ||
Set Membership: |
|