Search
Search

Triqk| Gene Symbol: Triqk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850092.2 | XP_004850149.1 | 2 - 3 | 222 | 91 | |
| XM_004850093.2 | XP_004850150.1 | 3 - 4 | 383 | 91 | ||
| XM_021262626.1 | XP_021118285.1 | 3 - 4 | 657 | 91 | ||
| XM_021262627.1 | XP_021118286.1 | 3 - 4 | 380 | 91 | ||
Set Membership: |
|