Search
Search

Psen2| Gene Symbol: Psen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853521.3 | XP_004853578.1 | 5 - 6 | 841 | 66 | |
| XM_004853522.3 | XP_004853579.1 | 5 - 6 | 842 | 66 | ||
| XM_021265544.1 | XP_021121203.1 | 5 - 6 | 693 | 66 | ||
| XM_021265545.1 | XP_021121204.1 | 5 - 6 | 842 | 66 | ||
| XM_021265546.1 | XP_021121205.1 | 5 - 6 | 843 | 66 | ||
| XM_021265547.1 | XP_021121206.1 | 5 - 6 | 842 | 66 | ||
| XM_021265548.1 | XP_021121207.1 | 5 - 6 | 844 | 66 | ||
Set Membership: |
|