Search
Search

Edrf1| Gene Symbol: Edrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838747.2 | XP_004838804.1 | 1 - 2 | 271 | 59 | |
| XM_004838748.2 | XP_004838805.1 | 1 - 2 | 271 | 59 | ||
| XM_004838749.2 | XP_004838806.1 | 1 - 2 | 271 | 59 | ||
| XM_004838750.2 | XP_004838807.1 | 1 - 2 | 271 | 59 | ||
Set Membership: |
|