Search
Search

Pear1| Gene Symbol: Pear1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871262.3 | XP_004871319.1 | 14 - 15 | 2156 | 66 | |
| XM_013067966.2 | XP_012923420.1 | 20 - 21 | 2954 | 66 | ||
| XM_013067967.2 | XP_012923421.1 | 14 - 15 | 2126 | 66 | ||
| XM_021249368.1 | XP_021105027.1 | 19 - 20 | 5525 | 66 | ||
| XM_021249369.1 | XP_021105028.1 | 19 - 20 | 4209 | 66 | ||
Set Membership: |
|