Search
Search

Txndc9| Gene Symbol: Txndc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844428.3 | XP_004844485.1 | 4 - 5 | 805 | 57 | |
| XM_021258070.1 | XP_021113729.1 | 4 - 5 | 853 | 57 | ||
| XM_021258071.1 | XP_021113730.1 | 4 - 5 | 673 | 57 | ||
Set Membership: |
|