Search
Search

Agtrap| Gene Symbol: Agtrap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863568.2 | XP_004863625.1 | 3 - 4 | 290 | 86 | |
| XM_021243555.1 | XP_021099214.1 | 2 - 3 | 230 | 86 | ||
Set Membership: |
|