Search
Search

Col4a1| Gene Symbol: Col4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858556.3 | XP_004858613.1 | 22 - 23 | 1445 | 58 | |
| XM_021239361.1 | XP_021095020.1 | 22 - 23 | 1332 | 58 | ||
| XM_021239362.1 | XP_021095021.1 | 22 - 23 | 1362 | 58 | ||
Set Membership: |
|