Search
Search

Ccdc30| Gene Symbol: Ccdc30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871919.3 | XP_004871976.2 | 10 - 11 | 1827 | 71 | |
| XM_021249924.1 | XP_021105583.1 | 7 - 8 | 1202 | 71 | ||
| XM_021249925.1 | XP_021105584.1 | 10 - 11 | 1826 | 71 | ||
| XM_021249927.1 | XP_021105586.1 | 10 - 11 | 1826 | 71 | ||
| XM_021249928.1 | XP_021105587.1 | 10 - 11 | 1825 | 71 | ||
Set Membership: |
|