Search
Search

Tmem245| Gene Symbol: Tmem245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073437.2 | XP_012928891.1 | 4 - 5 | 933 | 68 | |
| XM_013073438.2 | XP_012928892.1 | 4 - 5 | 933 | 68 | ||
Set Membership: |
|