Search
Search

Slc26a5| Gene Symbol: Slc26a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839711.2 | XP_004839768.1 | 6 - 7 | 5338 | 62 | |
| XM_004839712.2 | XP_004839769.1 | 6 - 7 | 5338 | 62 | ||
| XM_021253950.1 | XP_021109609.1 | 6 - 7 | 899 | 62 | ||
| XM_021253951.1 | XP_021109610.1 | 5 - 6 | 755 | 62 | ||
Set Membership: |
|