Search
Search

Col12a1| Gene Symbol: Col12a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013065253.2 | XP_012920707.1 | 46 - 47 | 7815 | 59 | |
| XM_013065254.2 | XP_012920708.1 | 46 - 47 | 7814 | 59 | ||
| XM_021243751.1 | XP_021099410.1 | 31 - 32 | 4311 | 59 | ||
Set Membership: |
|