Search
Search

Phldb2| Gene Symbol: Phldb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835746.2 | XP_004835803.1 | 4 - 5 | 1993 | 69 | |
| XM_004835747.2 | XP_004835804.1 | 4 - 5 | 1993 | 69 | ||
| XM_004835748.2 | XP_004835805.1 | 4 - 5 | 1993 | 69 | ||
| XM_004835749.2 | XP_004835806.1 | 4 - 5 | 1993 | 69 | ||
| XM_004835750.2 | XP_004835807.1 | 4 - 5 | 1993 | 69 | ||
| XM_021263804.1 | XP_021119463.1 | 4 - 5 | 1994 | 69 | ||
| XM_021263808.1 | XP_021119467.1 | 4 - 5 | 1993 | 69 | ||
| XM_021263815.1 | XP_021119474.1 | 3 - 4 | 1812 | 69 | ||
| XM_021263822.1 | XP_021119481.1 | 3 - 4 | 1886 | 69 | ||
| XM_021263828.1 | XP_021119487.1 | 3 - 4 | 1769 | 69 | ||
| XM_021263832.1 | XP_021119491.1 | 3 - 4 | 1911 | 69 | ||
| XM_021263834.1 | XP_021119493.1 | 3 - 4 | 1814 | 69 | ||
| XM_021263835.1 | XP_021119494.1 | 3 - 4 | 1873 | 69 | ||
| XM_021263838.1 | XP_021119497.1 | 4 - 5 | 1993 | 69 | ||
Set Membership: |
|