Search
Search

Stk32c| Gene Symbol: Stk32c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838712.2 | XP_004838769.1 | 4 - 5 | 827 | 63 | |
| XM_021253249.1 | XP_021108908.1 | 4 - 5 | 828 | 63 | ||
| XM_021253250.1 | XP_021108909.1 | 4 - 5 | 829 | 63 | ||
| XM_021253251.1 | XP_021108910.1 | 4 - 5 | 830 | 63 | ||
| XM_021253252.1 | XP_021108911.1 | 3 - 4 | 531 | 63 | ||
| XM_021253253.1 | XP_021108912.1 | 4 - 5 | 833 | 63 | ||
Set Membership: |
|