Search
Search

LOC101718059| Gene Symbol: LOC101718059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078064.2 | XP_012933518.1 | 2 - 3 | 1701 | 77 | |
| XM_013078069.2 | XP_012933523.1 | 2 - 3 | 1412 | 77 | ||
| XM_021240817.1 | XP_021096476.1 | 2 - 3 | 1748 | 77 | ||
| XM_021240818.1 | XP_021096477.1 | 2 - 3 | 1727 | 77 | ||
| XM_021240819.1 | XP_021096478.1 | 3 - 4 | 1477 | 77 | ||
| XM_021240820.1 | XP_021096479.1 | 2 - 3 | 1748 | 77 | ||
Set Membership: |
|